Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HX 630, Tocris Bioscience™

Catalog No. 391350
Change view
Click to view available options
Quantity:
10 mg
50 mg
Catalog No. Quantity
39-135-0 50 mg
39-131-0 10 mg
2 options
This item is not returnable. View return policy

Catalog No. 39-135-0

Supplier: R&D Systems 3913/50

This item is not returnable. View return policy

RXR synergist

RXR agonist. Acts as a retinoid synergist; enhances the potency of AM 80 (Cat. No. 3507) in a HL-60 cell differentiation assay.

Chemical Identifiers

CAS 188844-52-2
Molecular Formula C28H27NO2S
Molecular Weight (g/mol) 441.589
InChI Key PFGCWQPTOKPRRK-UHFFFAOYSA-N
Synonym 4-7,8,9,10-tetrahydro-7,7,10,10-tetramethylbenzo b naphtho 2,3-f 1,4 thiazepin-12-yl-benzoic acid, benzoic acid, 4-7,8,9,10-tetrahydro-7,7,10,10-tetramethylbenzo b naphtho 2,3-f 1,4 thiazepin-12-yl, 4-7,7,10,10-tetramethyl-7,8,9,10-tetrahydro-5-thia-13-aza-benzo 4,5 cyclohepta 1,2-b naphthalen-12-yl-benzoic acid, 4-7,8,9,10-tetrahydro-7,7,10,10-tetramethylbenzo b naphtho 2,3-f 1,4 thiazepin-12-yl benzoic acid, 4-14,14,17,17-tetramethyl-2-thia-9-azatetracyclo 9.8.0.0 3 ,?.0 1 3 , 1 ? nonadeca-1 19 ,3,5,7,9,11,13 18-heptaen-10-yl benzoic acid
PubChem CID 9889522
IUPAC Name 4-(7,7,10,10-tetramethyl-8,9-dihydronaphtho[2,3-b][1,5]benzothiazepin-12-yl)benzoic acid
SMILES CC1(CCC(C2=C1C=C3C(=C2)SC4=CC=CC=C4N=C3C5=CC=C(C=C5)C(=O)O)(C)C)C

Specifications

Quantity 50 mg
Formula Weight 441.58
Percent Purity >98%
Chemical Name or Material HX 630
Recommended Storage Store at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.