Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mettler Toledo™ HX/HS Anti-Theft Device

Catalog No. 01912581 Shop All METTLER TOLEDO Products
Change view
Click to view available options
18 in.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
01-912-581 18 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-912-581 Supplier Mettler Toledo™ Supplier No. 11600361
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Strong coated steel cord with a loop at one end and a detachable lock with T-bar mechanism at the other.

  • Steel security cord
  • Detachable lock with T-bar mechanism
  • Visual and physical safety
  • Two keys provided
  • Anti-theft Solution
  • Easy Installation
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Equipment) Balances and Moisture Analyzers
Material Steel
Length (English) 18 in.
Length (Metric) 45 cm
For Use With (Application) Lock balances and moisture analyzers
Quantity 1
Weight (Metric) 220 g
Product Line Balance and Moisture Analyzer Accessory
Type Moisture Analyzer Accessory
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.