Learn More
Medchemexpress LLC HXR9 hydrochloride | 99.5% | 2754.67 | 1 MG
SDP
Catalog No. 5000366113
Encompass_Preferred
HXR9 hydrochloride is a cell-permeable peptide that acts as a competitive antagonist of HOX/PBX interaction. It selectively decreases cell proliferation and promotes apoptosis in cells with high HOXA/PBX3 gene expression, such as MLL-rearranged leukemic cells, by blocking the interaction between HOX and PBX transcription factors.
  • Antagonizes the interaction between HOX and PBX transcription factors.
  • Selectively decreases cell proliferation.
  • Promotes apoptosis in cells with high HOXA/PBX3 gene expression.
  • Appears as a white to off-white solid.
  • Soluble in water (H2O) and PBS.
Catalog No. 50-003-66113 Supplier Medchemexpress LLC Supplier No. 1MGHYP3245A
Add to Cart
Add to Cart
HXR9 hydrochloride is a cell-permeable peptide that acts as a competitive antagonist of HOX/PBX interaction. It selectively decreases cell proliferation and promotes apoptosis in cells with high HOXA/PBX3 gene expression, such as MLL-rearranged leukemic cells, by blocking the interaction between HOX and PBX transcription factors.
  • Antagonizes the interaction between HOX and PBX transcription factors.
  • Selectively decreases cell proliferation.
  • Promotes apoptosis in cells with high HOXA/PBX3 gene expression.
  • Appears as a white to off-white solid.
  • Soluble in water (H2O) and PBS.