Learn More
Medchemexpress LLC Hxr9 hydrochloride | 99.5% | 2754.67 | 5 MG
SDP
Catalog No. 5000398840
Encompass_Preferred
HXR9 hydrochloride is a cell-permeable peptide and a competitive antagonist of HOX/PBX interaction. It antagonizes the interaction between HOX and PBX, a transcription factor that binds to HOX proteins in paralogue groups 1 to 8. It selectively decreases cell proliferation and promotes apoptosis in cells with high HOXA/PBX3 genes expression, such as MLL-rearranged leukemic cells.
  • Cell-permeable peptide
  • Competitive antagonist of HOX/PBX interaction
  • Decreases cell proliferation in cells with high HOXA/PBX3 genes expression
  • Promotes apoptosis in cells with high HOXA/PBX3 genes expression
  • Blocks the interaction between PBX and HOX
  • Exhibits antiproliferative activity
  • Blocks tumor growth in vivo
Catalog No. 50-003-98840 Supplier Medchemexpress LLC Supplier No. HYP3245A5MG
HXR9 hydrochloride is a cell-permeable peptide and a competitive antagonist of HOX/PBX interaction. It antagonizes the interaction between HOX and PBX, a transcription factor that binds to HOX proteins in paralogue groups 1 to 8. It selectively decreases cell proliferation and promotes apoptosis in cells with high HOXA/PBX3 genes expression, such as MLL-rearranged leukemic cells.
  • Cell-permeable peptide
  • Competitive antagonist of HOX/PBX interaction
  • Decreases cell proliferation in cells with high HOXA/PBX3 genes expression
  • Promotes apoptosis in cells with high HOXA/PBX3 genes expression
  • Blocks the interaction between PBX and HOX
  • Exhibits antiproliferative activity
  • Blocks tumor growth in vivo