Learn More
Medchemexpress LLC Hy-078020 (compound III-4) | 2756222-90-7 | 98.9% | 515.09 g/mol | C32H35ClN2O2 | 10 MG
SDP
Catalog No. 5000288801
Encompass_Preferred
This small-molecule compound is a selective, orally active antagonist of the histamine H1 receptor with reported in vitro potency (H1 IC50 = 24.12 nM). It is supplied for research use with defined purity and storage recommendations.
  • Selective, orally active histamine H1 receptor antagonist (H1 IC50 = 24.12 nM).
  • Demonstrates selectivity versus muscarinic receptor (mAChR3 IC50 > 10 μM).
  • High purity (98.9%) as supplied.
  • Available as 10 mg powder and 10 mM solution in DMSO (1 mL).
  • Storage: powder -20°C (up to 3 years) or 4°C (up to 2 years); in solvent -80°C (up to 6 months).
  • Suitable for in vitro pharmacology and allergic disease research models.
Catalog No. 50-002-88801 Supplier Medchemexpress LLC Supplier No. HY16223110MG
This small-molecule compound is a selective, orally active antagonist of the histamine H1 receptor with reported in vitro potency (H1 IC50 = 24.12 nM). It is supplied for research use with defined purity and storage recommendations.
  • Selective, orally active histamine H1 receptor antagonist (H1 IC50 = 24.12 nM).
  • Demonstrates selectivity versus muscarinic receptor (mAChR3 IC50 > 10 μM).
  • High purity (98.9%) as supplied.
  • Available as 10 mg powder and 10 mM solution in DMSO (1 mL).
  • Storage: powder -20°C (up to 3 years) or 4°C (up to 2 years); in solvent -80°C (up to 6 months).
  • Suitable for in vitro pharmacology and allergic disease research models.