Learn More
STEMCELL Technologies ClonaCell™-HY Liquid HAT Selection Medium, Size: 500 mL
SDP
Catalog No. NC0791135
Encompass
ClonaCell™-HY Liquid HAT Selection Medium is a serum-containing medium with hypoxanthine, aminopterin, and thymidine (HAT) for the selection of hybridomas following the fusion of lymphocytes and myeloma cells. This medium has been verified for use in mouse and rat hydridoma development and reportedly is compatible for production and cloning of myelomas and/or hybridomas using lymphocytes from a variety of host animals including human, mouse, rat, and hamster.
Catalog No. NC0791135 Supplier STEMCELL Technologies Supplier No. 03831
ClonaCell™-HY Liquid HAT Selection Medium is a serum-containing medium with hypoxanthine, aminopterin, and thymidine (HAT) for the selection of hybridomas following the fusion of lymphocytes and myeloma cells. This medium has been verified for use in mouse and rat hydridoma development and reportedly is compatible for production and cloning of myelomas and/or hybridomas using lymphocytes from a variety of host animals including human, mouse, rat, and hamster.