Learn More
Waters Corp Waters Inlet Block, Hy-REF, Houses Reference Electrode
Catalog No. 50792665 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The Hy-REF Inlet Block is an essential component of a flowcell system. It is specifically designed to house the Hy-REF reference electrode, working in conjunction with a separate working electrode block and a spacer to form a complete flowcell.
  • Houses the reference electrode
  • Forms part of a flowcell assembly
  • Separated from the working electrode block by a spacer
Catalog No. 50-792-665 Supplier Waters Corp Supplier No. 700001965
The Hy-REF Inlet Block is an essential component of a flowcell system. It is specifically designed to house the Hy-REF reference electrode, working in conjunction with a separate working electrode block and a spacer to form a complete flowcell.
  • Houses the reference electrode
  • Forms part of a flowcell assembly
  • Separated from the working electrode block by a spacer