Learn More
Aviva Systems Biology Corp HYAL1 Antibody - N-terminal region (ARP50813_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP50813_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against HYAL1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human HYAL1. Alternate names: HYAL-1, LUCA1, MGC45987, NAT6ApplicationWB, IHC. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-60442