Learn More
Aviva Systems Biology Corp HYAL3 Antibody - C-terminal region (ARP61904_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP61904_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against HYAL3. It was validated on Western Blot. . Alternate names: LUCA-3, LUCA14, LUCA3, Minna14, HYAL-3ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Sheep
Catalog No. 50-287-502