Learn More
Medchemexpress LLC Hyaluronan-binding peptide, biotin labeled | 2012.25 | 1 MG
SDP
Catalog No. 5000398911
Encompass_Preferred
Hyaluronan-binding peptide, biotin labeled is a biologically active peptide. This peptide is biotinylated through a C-terminal GGGSK linker and binds to hyaluronan (HA). HA is a nonsulfated glycosaminoglycan found in the extracellular matrix and on cell surfaces, playing roles in various biological processes including fertilization, wound healing, and immune responses. The peptide can block HA binding to CD44 receptors and inhibit T cell proliferation.
  • Biologically active peptide
  • Biotinylated through a C-terminal GGGSK linker
  • Binds to hyaluronan (HA)
  • Can block HA binding to CD44 receptors
  • Inhibits T cell proliferation
Catalog No. 50-003-98911 Supplier Medchemexpress LLC Supplier No. HYP5351A1MG
Hyaluronan-binding peptide, biotin labeled is a biologically active peptide. This peptide is biotinylated through a C-terminal GGGSK linker and binds to hyaluronan (HA). HA is a nonsulfated glycosaminoglycan found in the extracellular matrix and on cell surfaces, playing roles in various biological processes including fertilization, wound healing, and immune responses. The peptide can block HA binding to CD44 receptors and inhibit T cell proliferation.
  • Biologically active peptide
  • Biotinylated through a C-terminal GGGSK linker
  • Binds to hyaluronan (HA)
  • Can block HA binding to CD44 receptors
  • Inhibits T cell proliferation