Learn More
Creative Pegworks Hyaluronate Biotin, 10 mg
SDP
Supplier:  Creative Pegworks HA60210MG
Encompass
Hyaluronic Acid is labeled with biotin. Purity: >95% lyophilized powder. Degree of substitution (DoS): 1 mol % substitution. Other biotin labeling ratio such as 5% can be ordered through custom synthesis.
Catalog No. NC1812797