Learn More
Worthington Biochemical Corporation HYALURONIC ACID 10MG
SDP
Supplier:  Worthington Biochemical Corporation LS003907
Encompass
A partially purified powder. Suitable as a substrate for hyaluronidase assays. 9004-61-9
Catalog No. 50-592-405