Learn More
Echelon Biosciences Research Labs Hyaluronan Enzyme-Linked Immunosorbent Assay (HA ELISA)
SDP
Supplier:  Echelon Biosciences Research Labs K1200EAKIT
Encompass
The Hyaluronan Enzyme-Linked Immunosorbent Assay (HA ELISA), a competitive 96-well assay, offers a simple, effective method for determining Hyaluronic Acid (HA) levels in human and animal biological fluids or cell supernatants.Assay Range: 50-1600 ng/mlSensitivity: 25 ng/mLSample considerations: The HA ELISA detects HA molecules that are as small as 6.4 kDA This item is non returnable
Catalog No. 50-720-423