Learn More
Biomatik Corporation Hyaluronic Acid (HA) Mini sample ELISA Kit
Supplier:  Biomatik Corporation EKU12121
Encompass_Preferred
Hyaluronic Acid (HA) Mini sample ELISA Kit for detecting HA in serum, plasma, other biological fluids. Species Reactivity: Universal. Detection Range: 4.94-400ng/ml. Sensitivity: 1.77ng/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 3h. Target Synonyms: Hyaluronan; Hyaluronate. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8025