Learn More
TARGETMOL CHEMICALS INC HYALURONIC ACID SODIUM 500MG
SDP
Catalog No. 502579825
Encompass_Preferred
Also available in 50 mg 100 mg 200 mg 1000 mg and bulk. Please contact Fisher for quotes. Hyaluronic acid sodium (Sodium Hyaluronate) is an anionic nonsulfated glycosaminoglycan distributed widely throughout connective epithelial and neural tissues. It is unique among glycosaminoglycans in that it is nonsulfated forms in the plasma membrane instead of the Golgi and can be very large with its molecular weight often reaching the millions. One of the chief components of the extracellular matrix hyaluronic acid contributes significantly to cell proliferation and migration and may also be involved in the progression of some malignant tumors. purity: 98%
Catalog No. 50-257-9825 Supplier TARGETMOL CHEMICALS INC Supplier No. T5004500MG
Also available in 50 mg 100 mg 200 mg 1000 mg and bulk. Please contact Fisher for quotes. Hyaluronic acid sodium (Sodium Hyaluronate) is an anionic nonsulfated glycosaminoglycan distributed widely throughout connective epithelial and neural tissues. It is unique among glycosaminoglycans in that it is nonsulfated forms in the plasma membrane instead of the Golgi and can be very large with its molecular weight often reaching the millions. One of the chief components of the extracellular matrix hyaluronic acid contributes significantly to cell proliferation and migration and may also be involved in the progression of some malignant tumors. purity: 98%