Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Hyaluronic acid (sodium) | 9067-32-7 | 98.0% | 1500 kDa | 100 MG
SDP

Catalog No. 5000364893
Encompass_Preferred

Hyaluronic acid (sodium) | 9067-32-7 | 98.0% | 1500 kDa | 100 MG

Catalog No. 50-003-64893 Supplier Medchemexpress LLC Supplier No. 100MGHYB0633
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Hyaluronic acid (sodium) | 9067-32-7 | 98.0% | 1500 kDa | 100 MG

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.