Learn More
Medchemexpress LLC Hyaluronic acid (sodium) | 9067-32-7 | 98.0% | 500 MG
SDP
Catalog No. 5000408906
Encompass_Preferred
Hyaluronic acid sodium (Sodium hyaluronate) is a biopolymer made of repeating disaccharide units. It is a main component of the extracellular matrix (ECM) and plays a significant role in various physiological processes, including wound healing, ovulation, fertilization, and signal transduction. It is also studied in the context of joint diseases and cancer.
  • Activates PI3K-Akt signaling and regulates cancer-associated lymphangiogenesis
  • Boosts cell invasion and angiogenesis by promoting proteolytic MMP-9 binding
  • Used as a drug delivery agent for sodium butyrate to enhance anti-proliferative activity
  • Improves tissue hydration and resistance to mechanical damage
  • Reduces chemotaxis and migration of inflammatory cells
Catalog No. 50-004-08906 Supplier Medchemexpress LLC Supplier No. HYB0633500MG
Hyaluronic acid sodium (Sodium hyaluronate) is a biopolymer made of repeating disaccharide units. It is a main component of the extracellular matrix (ECM) and plays a significant role in various physiological processes, including wound healing, ovulation, fertilization, and signal transduction. It is also studied in the context of joint diseases and cancer.
  • Activates PI3K-Akt signaling and regulates cancer-associated lymphangiogenesis
  • Boosts cell invasion and angiogenesis by promoting proteolytic MMP-9 binding
  • Used as a drug delivery agent for sodium butyrate to enhance anti-proliferative activity
  • Improves tissue hydration and resistance to mechanical damage
  • Reduces chemotaxis and migration of inflammatory cells