Learn More
Creative Pegworks HYALURONIC NHS ESTER 10K 500MG
SDP
Supplier:  Creative Pegworks HA3411G
Encompass
Hyaluronate NHS ester, MW 10k Hyaluronic Acid is functionalized with NHS ester groups. Purity: >95% powder. Degree of substitution: 5 mol % substitution
Catalog No. NC1571930