Learn More
Medchemexpress LLC HY-107910 10mg Medchemexpress, Hyaluronidase CAS:37326-33-3 Purity:>98%
SDP
Catalog No. 502080725
Encompass_Preferred
Medchemexpress, HY-107910 10mg Hyaluronidase CAS:37326-33-3 Hyaluronidase (Hyaluronate 4-glycanohydrolase; Hyaluronoglucosaminidase) is a naturally occurring enzyme that depolymerizes hyaluronic acid by cleavage of glycosidic bonds and has been utilized in ophthalmic surgery. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-0725 Supplier Medchemexpress LLC Supplier No. HY10791010MG
Medchemexpress, HY-107910 10mg Hyaluronidase CAS:37326-33-3 Hyaluronidase (Hyaluronate 4-glycanohydrolase; Hyaluronoglucosaminidase) is a naturally occurring enzyme that depolymerizes hyaluronic acid by cleavage of glycosidic bonds and has been utilized in ophthalmic surgery. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.