Learn More
Worthington Biochemical Corporation HYALURONIDASE 300KU
SDP
Catalog No. NC9992317
Encompass
Hyaluronidase, 300 ku/Pack, A partially purified, dialyzed, lyophilized powder, Store at -20degreeC, Source: Bovine Testes, Minimum Activity: =300 USP/NF units per mg dry weight
Catalog No. NC9992317 Supplier Worthington Biochemical Corporation Supplier No. LS002592
Hyaluronidase, 300 ku/Pack, A partially purified, dialyzed, lyophilized powder, Store at -20degreeC, Source: Bovine Testes, Minimum Activity: =300 USP/NF units per mg dry weight