Learn More
Medchemexpress LLC Hyaluronidase, bovine testes | 37326-33-3 | MFCD00131351 | 55,000 Da (average) | 5 MG
SDP
Catalog No. 5000242285
Encompass_Preferred
Hyaluronidase, bovine testes is an endoglycosidase that depolymerizes hyaluronic acid (HA) by cleaving glycosidic bonds. Supplied as a solid for research use, it is applied in tissue dissociation, studies of HA-mediated signaling, and research addressing HA filler complications.
  • Degrades hyaluronic acid for tissue dissociation assays.
  • Facilitates investigation of HA-related signaling pathways.
  • Applicable to research models of HA filler complications and tissue repair.
  • Typical average molecular weight of approximately 55,000 Da.
  • Provided in small research quantities suitable for experimental use.
Catalog No. 50-002-42285 Supplier Medchemexpress LLC Supplier No. HY1079105MG
Hyaluronidase, bovine testes is an endoglycosidase that depolymerizes hyaluronic acid (HA) by cleaving glycosidic bonds. Supplied as a solid for research use, it is applied in tissue dissociation, studies of HA-mediated signaling, and research addressing HA filler complications.
  • Degrades hyaluronic acid for tissue dissociation assays.
  • Facilitates investigation of HA-related signaling pathways.
  • Applicable to research models of HA filler complications and tissue repair.
  • Typical average molecular weight of approximately 55,000 Da.
  • Provided in small research quantities suitable for experimental use.