Learn More
Crescent Chemical Co Inc HYALURONIDASE FRM OVINE TESTES
SDP
Catalog No. 50247724
Encompass_Preferred
Hyaluronidase from ovine testes min. 1000 U/mg lyophil. Unit definition: 1 U produces the same turbidity reduction in a mixture of hyaluronic acid and albumin as 1 I.U. (International Unit) of a standard hyaluronidase preparation (1). Size - 500MG Storage Conditions - -15 °C TO -25 °C
Catalog No. 50-247-724 Supplier Crescent Chemical Co Inc Supplier No. SE25118.02
Hyaluronidase from ovine testes min. 1000 U/mg lyophil. Unit definition: 1 U produces the same turbidity reduction in a mixture of hyaluronic acid and albumin as 1 I.U. (International Unit) of a standard hyaluronidase preparation (1). Size - 500MG Storage Conditions - -15 °C TO -25 °C