Learn More
Sigma Aldrich Fine Chemicals Biosciences Hyaluronidase from sheep testes, 37326-33-3, MFCD00131351, 1 g
Catalog No. NC1617570
Encompass
Type II, lyophilized powder, ≥300 units/mg solid, storage temp: −20°C, Synonym: Hyaluronate 4-glycanohydrolase, Hyaluronoglucosaminidase.
Catalog No. NC1617570 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. H21261G
Type II, lyophilized powder, ≥300 units/mg solid, storage temp: −20°C, Synonym: Hyaluronate 4-glycanohydrolase, Hyaluronoglucosaminidase.