Learn More
Medchemexpress LLC Hyaluronidase, ovine testes | 37326-33-3 | MFCD00131351 | 95.0% | 55,000 g/mol | 5 MG
SDP
Catalog No. 5000263681
Encompass_Preferred
Hyaluronidase, ovine testes is a protein enzyme preparation that hydrolyzes hyaluronic acid in extracellular matrices. Supplied as a lyophilized powder, it is intended for use in biochemical assays, tissue dissociation, and reproductive biology research where controlled degradation of hyaluronic acid is required.
  • Enzymatic degradation of hyaluronic acid for tissue dissociation and analytic assays.
  • Derived from ovine testes for endogenous enzyme activity profile.
  • High purity to minimize contaminating proteins and activities (95.0%).
  • Average molecular weight approximately 55,000 g/mol for preparative considerations.
  • Supplied in small pack sizes suitable for research-scale experiments.
Catalog No. 50-002-63681 Supplier Medchemexpress LLC Supplier No. HY108903A5MG
Hyaluronidase, ovine testes is a protein enzyme preparation that hydrolyzes hyaluronic acid in extracellular matrices. Supplied as a lyophilized powder, it is intended for use in biochemical assays, tissue dissociation, and reproductive biology research where controlled degradation of hyaluronic acid is required.
  • Enzymatic degradation of hyaluronic acid for tissue dissociation and analytic assays.
  • Derived from ovine testes for endogenous enzyme activity profile.
  • High purity to minimize contaminating proteins and activities (95.0%).
  • Average molecular weight approximately 55,000 g/mol for preparative considerations.
  • Supplied in small pack sizes suitable for research-scale experiments.