Learn More
NEXPRING HEALTH Hyaluronidase Solution, 5 x 1 mL
Catalog No. NC0578732
Encompass
For the removal of cumulus cells surrounding freshly retrieved oocytes.
Catalog No. NC0578732 Supplier NEXPRING HEALTH Supplier No. 901015X1ML
For the removal of cumulus cells surrounding freshly retrieved oocytes.