Learn More
Supelco Inc HYALURONIDASE TYPE 1V-S
Supplier:  Supelco Inc H3884100MG
Encompass
Hyaluronidase, Type IV-S, lyophilized powder (essentially salt-free), 750-3000A units/mg solid
Catalog No. NC9725814