Learn More
Hyamine 1622, 0.004 Molar (N) (Benzethonium Chloride Solution), Reagents
Hyamine 1622, 0.004 Molar (N) (Benzethonium Chloride Solution), Reagent, for general laboratory use. Manufactured in ISO 9001 facility.
$133.00 - $338.00
Specifications
| Chemical Name or Material | Hyamine 1622, 0.004 Molar (N) (Benzethonium Chloride Solution) |
|---|---|
| Color | Colorless |
| CAS | 7732-18-5 , 121-54-0 |
| Boiling Point | 100°C |
| Synonym | Benzethonium chloride |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|
01-335-065
|
Reagents Holdings Llc
CH11470010F |
10 L | Cubitainer |
Each for $338.00
|
|
||||
|
01-335-063
|
Reagents Holdings Llc
CH1147004C |
4 L | Glass Bottle |
Each for $197.00
|
|
||||
|
01-335-062
|
Reagents Holdings Llc
CH114700500A |
500 mL | Poly Bottle |
Each for $133.00
|
|
||||
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.