CAS RN 121-54-0
Benzethonium chloride, 97%
CAS: 121-54-0 Molecular Formula: C27H42ClNO2 Molecular Weight (g/mol): 448.08 MDL Number: MFCD00011742 InChI Key: UREZNYTWGJKWBI-UHFFFAOYSA-M Synonym: benzethonium chloride,quatrachlor,hyamine,phemeride,phemerol chloride,benzethoniumchloride,phemithyn,disilyn,kylacol,solamin PubChem CID: 8478 ChEBI: CHEBI:31264 IUPAC Name: benzyl-dimethyl-[2-[2-[4-(2,4,4-trimethylpentan-2-yl)phenoxy]ethoxy]ethyl]azanium;chloride SMILES: CC(C)(C)CC(C)(C)C1=CC=C(C=C1)OCCOCC[N+](C)(C)CC2=CC=CC=C2.[Cl-]
Hyamine Solution, 0.004 M (0.004 N), Ricca Chemical
CAS: 121-54-0 Molecular Formula: C27H42ClNO2 Molecular Weight (g/mol): 448.088 InChI Key: UREZNYTWGJKWBI-UHFFFAOYSA-M Synonym: benzethonium chloride,quatrachlor,hyamine,phemeride,phemerol chloride,benzethoniumchloride,phemithyn,disilyn,kylacol,solamin PubChem CID: 8478 ChEBI: CHEBI:31264 IUPAC Name: benzyl-dimethyl-[2-[2-[4-(2,4,4-trimethylpentan-2-yl)phenoxy]ethoxy]ethyl]azanium;chloride SMILES: CC(C)(C)CC(C)(C)C1=CC=C(C=C1)OCCOCC[N+](C)(C)CC2=CC=CC=C2.[Cl-]
Benzethonium Chloride 97.0+%, TCI America™
CAS: 121-54-0 Molecular Formula: C27H42ClNO2 Molecular Weight (g/mol): 448.088 MDL Number: MFCD00011742 InChI Key: UREZNYTWGJKWBI-UHFFFAOYSA-M Synonym: benzethonium chloride,quatrachlor,hyamine,phemeride,phemerol chloride,benzethoniumchloride,phemithyn,disilyn,kylacol,solamin PubChem CID: 8478 ChEBI: CHEBI:31264 IUPAC Name: benzyl-dimethyl-[2-[2-[4-(2,4,4-trimethylpentan-2-yl)phenoxy]ethoxy]ethyl]azanium;chloride SMILES: CC(C)(C)CC(C)(C)C1=CC=C(C=C1)OCCOCC[N+](C)(C)CC2=CC=CC=C2.[Cl-]
Benzethonium Chloride, USP, 97-103%, Spectrum™ Chemical
CAS: 121-54-0 Molecular Formula: C27H42ClNO2 Molecular Weight (g/mol): 448.09 InChI Key: UREZNYTWGJKWBI-UHFFFAOYSA-M IUPAC Name: benzyldimethyl(2-{2-[4-(2,4,4-trimethylpentan-2-yl)phenoxy]ethoxy}ethyl)azanium chloride SMILES: [Cl-].CC(C)(C)CC(C)(C)C1=CC=C(OCCOCC[N+](C)(C)CC2=CC=CC=C2)C=C1
Benzethonium chloride, 97%
CAS: 121-54-0 Molecular Formula: C27H42ClNO2 MDL Number: MFCD00011742 InChI Key: UREZNYTWGJKWBI-UHFFFAOYSA-M Synonym: benzethonium chloride,quatrachlor,hyamine,phemeride,phemerol chloride,benzethoniumchloride,phemithyn,disilyn,kylacol,solamin PubChem CID: 8478 ChEBI: CHEBI:31264
Benzethonium Chloride, 0.004 M (N), Ricca Chemical
CAS: 121-54-0 Molecular Formula: C27H42ClNO2 Molecular Weight (g/mol): 448.088 InChI Key: UREZNYTWGJKWBI-UHFFFAOYSA-M Synonym: benzethonium chloride,quatrachlor,hyamine,phemeride,phemerol chloride,benzethoniumchloride,phemithyn,disilyn,kylacol,solamin PubChem CID: 8478 ChEBI: CHEBI:31264 IUPAC Name: benzyl-dimethyl-[2-[2-[4-(2,4,4-trimethylpentan-2-yl)phenoxy]ethoxy]ethyl]azanium;chloride SMILES: CC(C)(C)CC(C)(C)C1=CC=C(C=C1)OCCOCC[N+](C)(C)CC2=CC=CC=C2.[Cl-]
Sigma Aldrich Hyamine™ 1622 Solution
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
Ricca Chemical Company Benzethonium Chloride, 0.004 Molar (Normal), Ricca Chemical
CAS: 121-54-0 Molecular Formula: C27H42ClNO2 Molecular Weight (g/mol): 448.088 InChI Key: UREZNYTWGJKWBI-UHFFFAOYSA-M Synonym: benzethonium chloride,quatrachlor,hyamine,phemeride,phemerol chloride,benzethoniumchloride,phemithyn,disilyn,kylacol,solamin PubChem CID: 8478 ChEBI: CHEBI:31264 IUPAC Name: benzyl-dimethyl-[2-[2-[4-(2,4,4-trimethylpentan-2-yl)phenoxy]ethoxy]ethyl]azanium;chloride SMILES: CC(C)(C)CC(C)(C)C1=CC=C(C=C1)OCCOCC[N+](C)(C)CC2=CC=CC=C2.[Cl-]