Learn More
Hyamine 1622, 0.004 Molar (N) (Benzethonium Chloride Solution), Reagents
Hyamine 1622, 0.004 Molar (N) (Benzethonium Chloride Solution), Reagent, for general laboratory use. Manufactured in ISO 9001 facility.
Supplier:  Reagents Holdings Llc CH1147004C
 View more versions of this product
Catalog No. 01-335-063
 Restricted Item
Add to Cart
Description
Description
Specifications
Specifications
Hyamine 1622, 0.004 Molar (N) (Benzethonium Chloride Solution)
7732-18-5 , 121-54-0
Benzethonium chloride
Glass Bottle
4 L
Liquid
Colorless
100°C
Reagent
1.001
Non-sterile
Product Suggestions
Product Suggestions