Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Hyamine™ 1622 Solution

Catalog No. 511265LF Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
1 L
5 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
51-126-5LF 5 L
51-126-1LF 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 51-126-5LF Supplier Sigma Aldrich Supplier No. 511265LF
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Hyamine™ 1622 is a cationic detergent benzethonium chloride. Poly(vinyl chloride) membrane ion-selective electrode for the Hyamine 1622 cation has been proposed. Its sorption on soil surfaces has been investigated. It is widely used in cosmetics and shampoos due to its antimicrobial and cationic surfactant properties.Hyamine™ 1622 solution was used as surfactant to examine its acute toxicity and effect on oxidative stress and cholinesterase (ChE) activity in Dugesia japonica. It may be employed as stopping reagent in the improved Eliman procedure for erythrocyte cholinesterase.Hyamine is a registered trademark of Lonza Group Ltd.

Specifications

CAS 121-54-0
Density 0.998 g/mL at 20°C
MDL Number MFCD00011742
Quantity 5 L
Synonym (Diisobutylphenoxyethoxyethyl)dimethylbenzylammonium chloride solution
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.