Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hyamine Solution, 0.004 Normal (0.004 Molar) (Benzethonium Chloride Solution), Reagents

Catalog No. 01335066
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-335-066 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-335-066 Supplier Reagents Holdings Llc Supplier No. CH1146001C
Add to Cart
Add to Cart

Hyamine Solution, 0.004 Normal (0.004 Molar) (Benzethonium Chloride Solution), Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Specifications

Chemical Name or Material Hyamine Solution, 0.004 Normal (0.004 Molar) (Benzethonium Chloride Solution)
Color Colorless
CAS 7732-18-5, 121-54-0
Boiling Point 100°C
Synonym Benzethonium chloride
Grade Reagent
Packaging Glass Bottle
Specific Gravity 1.001
Quantity 1 L
Sterility Non-sterile
Physical Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.