Learn More
Scigene Corporation HYBEX BLOCK 96X0.2ML TUBES
Catalog No. NC9556112
Encompass
Hybex Microtube heat block. Holds 96x0.2ml PCR tubes or single plate.
Catalog No. NC9556112 Supplier Scigene Corporation Supplier No. 1057310
Hybex Microtube heat block. Holds 96x0.2ml PCR tubes or single plate.