Learn More
Electron Microscopy Sciences hybex™ Media Storage Bottles Starter Pack, GL45 Cap, Yellow
SDP
Catalog No. 501930020
Encompass_Preferred
Reusable media bottles manufactured from high quality, borosilicate 3.3 glass. They feature superior chemical resistance and are ideal for the storage of reagents, culture media, biological fluids and many other aqueous and nonaqueous solutions.
All hybex bottles have permanent white enamel graduations for volume measurment, along with a white identification box for coding or marking.
With a low coefficient of thermal expansion (3.3), these bottles can withstand temperatures from -70 to 500 deg C. This allows for repeated use in freezers, microwaves, autoclaves and dry heat sterilization chambers.
Cap thread size: GL45
Color: Yellow
This item is not returnable. View return policy
Catalog No. 50-193-0020 Supplier Electron Microscopy Sciences Supplier No. 6425507Y
Reusable media bottles manufactured from high quality, borosilicate 3.3 glass. They feature superior chemical resistance and are ideal for the storage of reagents, culture media, biological fluids and many other aqueous and nonaqueous solutions.
All hybex bottles have permanent white enamel graduations for volume measurment, along with a white identification box for coding or marking.
With a low coefficient of thermal expansion (3.3), these bottles can withstand temperatures from -70 to 500 deg C. This allows for repeated use in freezers, microwaves, autoclaves and dry heat sterilization chambers.
Cap thread size: GL45
Color: Yellow