Learn More
Scigene Corporation HYBEX MICROSAMPLE INCUBATOR
Catalog No. NC9555274
Encompass
Hybex Microsample Incubator, base, 115V. A tube block, waterbath insert or microarray chamber kit is required for use of this instrument - purchase separately.
Catalog No. NC9555274 Supplier Scigene Corporation Supplier No. 1057300
Hybex Microsample Incubator, base, 115V. A tube block, waterbath insert or microarray chamber kit is required for use of this instrument - purchase separately.