Learn More
ATCC Hybri-Care Medium (Powder)
Catalog No. 502385108
Encompass
Hybri-Care Medium is formulated to support the growth of hybridomas and fastidious cell lines. A combined buffering system of HEPES and added NaHCO3 enables the medium to be used at all stages of hybridoma production from fusion to cloning, including single-cell subcloning. Each package contains dry powdered medium sufficient to produce one liter of 1X liquid medium. Net weight: 12 grams per package.
Catalog No. 50-238-5108 Supplier ATCC Supplier No. 46X
Add to Cart
Add to Cart
Hybri-Care Medium is formulated to support the growth of hybridomas and fastidious cell lines. A combined buffering system of HEPES and added NaHCO3 enables the medium to be used at all stages of hybridoma production from fusion to cloning, including single-cell subcloning. Each package contains dry powdered medium sufficient to produce one liter of 1X liquid medium. Net weight: 12 grams per package.