Learn More
Research Products International Corp Hybridization Bottle with Red Cap and PTFE Liner, Borosilicate Glass Bottles, 35 x 150 mm
SDP
Catalog No. 501978117
Encompass
Heavy wall borosilicate glass bottles designed for use in standard hybridization ovens. Fit all rotisseries that accommodate 35mm diameter bottles. Bottles feature a leakproof polypropylene closure with a PTFE/Silicone liner.
Catalog No. 50-197-8117 Supplier Research Products International Corp Supplier No. 246002
Heavy wall borosilicate glass bottles designed for use in standard hybridization ovens. Fit all rotisseries that accommodate 35mm diameter bottles. Bottles feature a leakproof polypropylene closure with a PTFE/Silicone liner.