Learn More
Mitegen Hybrid Grid Box with Pin Lid By Nanosoft, White, For Cryogenic Sample Organization & Protection
SDP
Catalog No. NC2100533
Encompass
This Hybrid Grid Box with Pin Lid by Nanosoft offers a versatile and secure solution for organized cryo-sample management. This listing features a single grid box in white. The hybrid grid box combines unique design elements to provide enhanced protection and ease of use for delicate samples under cryogenic conditions. The innovative pin lid ensures a precise and secure closure, actively safeguarding contents from contamination and physical damage during handling and storage. Its distinct white color aids in easy visual identification and categorization of samples, streamlining workflow efficiency and minimizing errors. This grid box is an essential tool for maintaining high standards in cryo-electron microscopy and other cryogenic research applications, where sample integrity is paramount. This grid box can be used in the MiTeGen Generation 2 Cryo-EM puck SKU M-CEMG2-1.
This item is not returnable. View return policy
Catalog No. NC2100533 Supplier Mitegen Supplier No. MCEMNSHGBPPL1W
This Hybrid Grid Box with Pin Lid by Nanosoft offers a versatile and secure solution for organized cryo-sample management. This listing features a single grid box in white. The hybrid grid box combines unique design elements to provide enhanced protection and ease of use for delicate samples under cryogenic conditions. The innovative pin lid ensures a precise and secure closure, actively safeguarding contents from contamination and physical damage during handling and storage. Its distinct white color aids in easy visual identification and categorization of samples, streamlining workflow efficiency and minimizing errors. This grid box is an essential tool for maintaining high standards in cryo-electron microscopy and other cryogenic research applications, where sample integrity is paramount. This grid box can be used in the MiTeGen Generation 2 Cryo-EM puck SKU M-CEMG2-1.