Learn More
Mitegen Hybrid Grid Box with Pin Lid by Nanosoft, Assorted Colors (12-Pack), For Cryogenic Sample Organization & Protection
SDP
Catalog No. NC2171347
Encompass
This Pack of 12 Hybrid Grid Boxes with Pin Lids by Nanosoft offers an excellent and versatile solution for organized and secure cryo-sample management in any laboratory. Each hybrid grid box combines unique design elements to provide enhanced protection and ease of use for delicate samples under cryogenic conditions. The innovative pin lid ensures a precise and secure closure, actively safeguarding contents from contamination and physical damage during handling and storage. These boxes are supplied in an assorted 12-pack, containing two of each vibrant color: White, Blue, Green, Purple, Yellow, and Pink. This diverse color-coding capability significantly enhances workflow efficiency and minimizes errors in high-throughput environments by facilitating easy visual identification and categorization of samples. These grid boxs can be used in the MiTeGen Generation 2 Cryo-EM puck SKU M-CEMG2-1.
This item is not returnable. View return policy
Catalog No. NC2171347 Supplier Mitegen Supplier No. MCEMNSHGBPPL12
This Pack of 12 Hybrid Grid Boxes with Pin Lids by Nanosoft offers an excellent and versatile solution for organized and secure cryo-sample management in any laboratory. Each hybrid grid box combines unique design elements to provide enhanced protection and ease of use for delicate samples under cryogenic conditions. The innovative pin lid ensures a precise and secure closure, actively safeguarding contents from contamination and physical damage during handling and storage. These boxes are supplied in an assorted 12-pack, containing two of each vibrant color: White, Blue, Green, Purple, Yellow, and Pink. This diverse color-coding capability significantly enhances workflow efficiency and minimizes errors in high-throughput environments by facilitating easy visual identification and categorization of samples. These grid boxs can be used in the MiTeGen Generation 2 Cryo-EM puck SKU M-CEMG2-1.