Learn More
Med Vet International Hybrid "T" Plates - 1.5mm or 2.0mm - Long, J0829B, Each
Supplier: Med Vet International JOR829B
The problem with many distal radius fractures is the distal segment is so small that it is difficult to place sufficient screws. A "T" plate can help but often the head of the plate is too wide. Hybrid "T" plates can provide a solution. - Screw holes in the head are smaller and closer together than those in the shaft - Plate will accept same size screw in the head as in the shaft, but with reduced screw angulations. Hybrid "T" Plate. 1.5 or 2mm head 2mm shaft 50mm length of shaft J-829a and J-829b are especially suitable for distal radius fractures in cats and miniature dogs.