Learn More
Med Vet International Hybrid "T" Plates 2.7mm Or 3.5mm, J0829, Each
Catalog No. 5000581965
Encompass_Preferred
The problem with many distal radius fractures is the distal segment is so small that it is difficult to place sufficient screws. A "T" plate can help but often the head of the plate is too wide. Hybrid "T" plates can provide a solution.Screw holes in the head are smaller and closer together than those in the shaftPlate will accept same size screw in the head as in the shaft, but with reduced screw angulations
Catalog No. 50-005-81965 Supplier Med Vet International Supplier No. JOR829
The problem with many distal radius fractures is the distal segment is so small that it is difficult to place sufficient screws. A "T" plate can help but often the head of the plate is too wide. Hybrid "T" plates can provide a solution.Screw holes in the head are smaller and closer together than those in the shaftPlate will accept same size screw in the head as in the shaft, but with reduced screw angulations