Learn More
LGC GENOMICS LLC (Lucigen) Hybridase Thermostable RNase H, 500 U @ 5 U/L
Catalog No. NC1229183
Encompass
Hybridase™ Thermostable RNase H specifically degrades the RNA in a DNA:RNA hybrid, without affecting DNA or unhybridized RNA.
This item is not returnable. View return policy
Catalog No. NC1229183 Supplier LGC GENOMICS LLC Supplier No. H39500
Hybridase™ Thermostable RNase H specifically degrades the RNA in a DNA:RNA hybrid, without affecting DNA or unhybridized RNA.