Learn More
Bioworld Hybridization Bags 404 (203x305mm) 40 bags/pk
Catalog No. NC9969061
Encompass_Preferred
Constructed of clear, 2.5mm polyester and polyethylene. These tough, durable bags can sustain temperatures from -56 to 115°C (-70 to 240°F). These transparent, airtight, and leak-proof pouches can be sterilized with gamma, infrared, and e-beam, and are easily sealed using a heat sealer. Once sealed, these pouches can also be surface sterilized using VHP. They are ideal for nonradioactive hybridization and detection procedures, standard radioactive probe hybridizations, and western blotting procedures. Specifications: 40 bags/PK, 203mm (W) X 305mm (L)
Catalog No. NC9969061 Supplier Bioworld Supplier No. 430110621
Constructed of clear, 2.5mm polyester and polyethylene. These tough, durable bags can sustain temperatures from -56 to 115°C (-70 to 240°F). These transparent, airtight, and leak-proof pouches can be sterilized with gamma, infrared, and e-beam, and are easily sealed using a heat sealer. Once sealed, these pouches can also be surface sterilized using VHP. They are ideal for nonradioactive hybridization and detection procedures, standard radioactive probe hybridizations, and western blotting procedures. Specifications: 40 bags/PK, 203mm (W) X 305mm (L)