Learn More
Seracare Life Sciences Inc HYBRIDIZATION BAGS EA=50BGS
Catalog No. 5067546
Encompass_Preferred
Hybridization bags are used for membrane incubation steps in Southern, Northern, and Western blotting protocols for such applications as hybridizations and washes, antibody and conjugate incubations, substrate application and film exposure. Hybridization Bags are 8 X 10 inch, 4 mil thick, clear polyethylene bags. They are heat-sealable, resistant to temperatures up to at least 65 C, and do not curl when cut.
This item is not returnable. View return policy
Catalog No. 50-675-46 Supplier Seracare Life Sciences Inc Supplier No. 59600026
Hybridization bags are used for membrane incubation steps in Southern, Northern, and Western blotting protocols for such applications as hybridizations and washes, antibody and conjugate incubations, substrate application and film exposure. Hybridization Bags are 8 X 10 inch, 4 mil thick, clear polyethylene bags. They are heat-sealable, resistant to temperatures up to at least 65 C, and do not curl when cut.