Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Electron Microscopy Sciences Secure-Seal™ Hybridization Chambers 1 well, 20mm 2.5mm deep
SDP

Catalog No. 5030445
Encompass_Preferred

SecureSeal adhesive chambers form peel-and-stick enclosures to isolate specimens affixed to glass microscope slides. RNase free, gasketed chambers provide optimum surface-to-volume fluid dynamics for hybridization assays on large or multiple specimens and genomic arrays on glass slides or film coated slides. Sealable access ports. Removes easily. All Hybridization Chambers have 1.5 mm diameter Ports and come with 120 Port Seals.
Wells: 1 well
I.D.: 20 mm
Depth: 2.5 mm
O.D.: 22 mm x 25 mm

Catalog No. 50-304-45 Supplier Electron Microscopy Sciences Supplier No. 7033336
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

SecureSeal adhesive chambers form peel-and-stick enclosures to isolate specimens affixed to glass microscope slides. RNase free, gasketed chambers provide optimum surface-to-volume fluid dynamics for hybridization assays on large or multiple specimens and genomic arrays on glass slides or film coated slides. Sealable access ports. Removes easily. All Hybridization Chambers have 1.5 mm diameter Ports and come with 120 Port Seals.
Wells: 1 well
I.D.: 20 mm
Depth: 2.5 mm
O.D.: 22 mm x 25 mm

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.