Learn More
Bioworld Hybridization Cocktail, 0% Formamide, 50 mL
Catalog No. 50196829
Encompass_Preferred
Hybridization Cocktail, 0% Formamide Speciffically designed to optimize complementary base-pairing of DNA and RNA in Southern Blot, Northern Blot, Dot Blot, Colony and in situhybridizations. The components of the Hybridization Cocktail will stabilize the target and probe nucleic acids, lower melting temperatures, concentrate the nucleic acids in solution and lower background on the membrane or slides. Ref: Unger E.R., Budgeon L.R., Meyerson D., Brigati D.J., Am. J. Surg. Pathol., 10(1): 1-8 (1986). 0% Formamide RNase/DNase: None Detected
Catalog No. 50-196-829 Supplier Bioworld Supplier No. 107500111
Add to Cart
Add to Cart
Hybridization Cocktail, 0% Formamide Speciffically designed to optimize complementary base-pairing of DNA and RNA in Southern Blot, Northern Blot, Dot Blot, Colony and in situhybridizations. The components of the Hybridization Cocktail will stabilize the target and probe nucleic acids, lower melting temperatures, concentrate the nucleic acids in solution and lower background on the membrane or slides. Ref: Unger E.R., Budgeon L.R., Meyerson D., Brigati D.J., Am. J. Surg. Pathol., 10(1): 1-8 (1986). 0% Formamide RNase/DNase: None Detected