Learn More
Bioworld Hybridization Cocktail, 45% Formamide, 50 mL
Supplier:  Bioworld 102500011
Encompass_Preferred
Hybridization Cocktail, 45% Formamide Designed to optimize complementary base-pairing of DNA and RNA in Southern Blot, Northern Blot, Dot Blot, Colony and in situ hybridizations. Components of these cocktails stabilize the target and probe nucleic acids, lower melting temperatures, concentrate the nucleic acids in solution and lower background on the membrane or slides. Reference: Unger E.R., Budgeon L.R., Meyerson D., Brigati D.J., Am. J. Surg. Pathol., 10(1): 1-8 (1986). Specifications:45% FormamideRNase/DNase: None Detected
Catalog No. 50-196-417