Learn More
Bioworld Hybridization Cocktail, 50% Formamide, 50 mL
Supplier:  Bioworld 107500121
Encompass_Preferred
Hybridization CocktailDesigned to optimize complementary base-pairing of DNA and RNA in Southern Blot, Northern Blot, Dot Blot, Colony and in situ hybridizations.Components of these cocktails stabilize the target and probe nucleic acids, lower melting temperatures, concentrate the nucleic acids in solution, and lower background on the membrane or slides. Specifications:50% Formamide50 mlStorage: -20° CRef: Unger E.R., Budgeon L.R., Meyerson D., Brigati D.J., Am. J. Surg. Pathol., 10(1): 1-8 (1986).
Catalog No. 50-196-831