Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Axygen™ HybridRack™ Pipet Tip, Extended Length, Maxymum Recovery Surface
GREENER_CHOICE

Catalog No. 01671155 Shop All Axygen Products
Change view
Click to view available options
Volume (Metric):
10 μL
300 μL
1000 μL
Color:
Blue
Green
Red
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) Color
01-671-155 10 μL Red
01-671-157 1000 μL Blue
01-671-149 300 μL Green
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 01-671-155 Supplier Axygen™ Supplier No. ERF10XTLRS
Add to Cart
Add to Cart

The HybridRack System removes approximately 70% of the plastic compared to traditional plastic pipet tip racks.

The Axygen HybridRack System combines sustainability and functionality to create a new, ecofriendly pipet tip product that is backed by Corning’s quality and value.

  • The HybridRack System removes approximately 70% of the plastic compared to traditional plastic pipet tip racks.
  • The plastic deck combined with the paperboard base provides a stable pipetting platform. The HybridRack is designed for end users to pipet directly from it, even when using a multi-channel instrument.
  • The paperboard base can be easily recycled through most local curbside recycling programs.
  • All HybridRack part numbers are provided sterile. Filtered, Maxymum Recovery, and non-filtered configurations are available.
  • The HybridRack System is manufactured in the USA at an ISO 13485, ISO 14001, and EnergyStar registered facility.
  • The pipet tips included in the HybridRack offering are the same tips as the Axygen MultiRack product line.
TRUSTED_SUSTAINABILITY

Specifications

Description Maxymum Recovery Surface
Volume (Metric) 10 μL
Filtered Filtered
Sterility Sterile
Color Red
Autoclavable Not Autoclavable
Format Rack
Tip Style Extended Length
Product Type Pipet Tip
Color Code Red
Disposable Single-use
For Use With (Equipment) Universal Fit
Graduated No
Material Polypropylene
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.