Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Axygen™ HybridRack™ Pipet Tip, Standard, Maxymum Recovery Surface
GREENER_CHOICE

Catalog No. 01671159 Shop All Axygen Products
Change view
Click to view available options
Volume (Metric):
20 μL
100 μL
200 μL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
01-671-159 100 μL
01-671-151 200 μL
01-671-153 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 01-671-159 Supplier Axygen™ Supplier No. ERF100LRS
Get Up to $2,500 in Products with Purchase
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The HybridRack System removes approximately 70% of the plastic compared to traditional plastic pipet tip racks.

The Axygen HybridRack System combines sustainability and functionality to create a new, ecofriendly pipet tip product that is backed by Corning’s quality and value.

  • The HybridRack System removes approximately 70% of the plastic compared to traditional plastic pipet tip racks.
  • The plastic deck combined with the paperboard base provides a stable pipetting platform. The HybridRack is designed for end users to pipet directly from it, even when using a multi-channel instrument.
  • The paperboard base can be easily recycled through most local curbside recycling programs.
  • All HybridRack part numbers are provided sterile. Filtered, Maxymum Recovery, and non-filtered configurations are available.
  • The HybridRack System is manufactured in the USA at an ISO 13485, ISO 14001, and EnergyStar registered facility.
  • The pipet tips included in the HybridRack offering are the same tips as the Axygen MultiRack product line.
TRUSTED_SUSTAINABILITY

Specifications

Description Maxymum Recovery Surface
Volume (Metric) 100 μL
Filtered Filtered
Sterility Sterile
Color Yellow
Autoclavable Not Autoclavable
Format Rack
Tip Style Standard
Product Type Pipet Tip
Disposable Single-use
For Use With (Equipment) Universal Fit
Graduated No
Material Polypropylene
Graduations None
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.