Learn More
MilliporeSigma™ Supelco™ HybridSPE™ DPX SPE Tip, Bed Wt. 30 mg
Catalog No. 111009773 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Volume (Metric):
200 μL
300 μL
1 mL
For Use With (Equipment):
Hamilton
INTEGRA
Tecan
Universal
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) For Use With (Equipment)
11-100-9773 200 μL Tecan
11-100-9774 300 μL Hamilton
11-100-9775 300 μL INTEGRA
11-100-9776 1 mL Universal
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 11-100-9773 Supplier MilliporeSigma™ Supelco™ Supplier No. 52973U
Add to Cart
Add to Cart
DPX stands for Dispersive Pipette EXtraction. HybridSPE DPX Tips are pipette tips that incorporate loosely contained HybridSPE sorbent material that is mixed with the sample solution to accomplish solid phase extraction.
HybridSPE technology is a simple and generic sample prep platform designed for the gross level removal of endogenous phospholipid interferences from biological plasma and serum prior to LC/MS or LC/MS/MS analysis. In this simple technique, biological plasma or serum is first subjected to protein precipitation via the addition and mixing of acidified acetonitrile. Precipitated proteins are then removed by centrifugation and the resulting supernatant is extracted using the HybridSPE DPX tip which acts as a chemical filter that specifically targets the removal of endogenous sample phospholipids. The phospholipid retention mechanism is based on a highly selective Lewis acid-base interaction between the proprietary zirconia ions functionally bonded to the HybridSPE stationary phase and the phosphate moiety consistent with all phospholipids. The resulting eluent is ready for immediate LC/MS or LC/MS/MS analysis.
MilliporeSigma Supelco offers an array of tips that are compatible with the most common liquid handling platforms and pipettors on the market: Tecan, Hamilton, INTEGRA, and a Universal format as well.
There are two versions of the DPX tips available for the HybridSPE-PL. The standard HybridSPE DPX 50 mg tips are designed for use with biological sample volumes between 100-300 muL, while the HybridSPE DPX 30 mg tips are designed for use with biological sample volumes between 30-100 muL.Guideline for 30 mg tips - plasma/serum sample: 30-100 muL; precipitating agent: 90-300 muL. Guideline for 50 mg tips - plasma/serum sample: 100-300 muL; precipitating agent: 300-900 muL.
  • Minimal elution solvent volumes.
  • Rapid extraction times (less than 3 minutes/sample).
  • High extraction efficiencies.
  • Easy to perform extractions.
  • Lower costs.
  • Higher throughput.
  • Minimal training required.
  • Environmentally friendly.
  • Eliminates ion suppression effects often associated with phospholipids.
  • Tips offered to fit many formats: Tecan, Hamilton, INTEGRA, and Universal liquid handling platforms and pipettors.
TRUSTED_SUSTAINABILITY
Specifications
Column Phase DPX
Column Type SPE
Bed Weight 30 mg
For Use With (Application) Food and Beverages
For Use With (Equipment) Tecan
Product Line HybridSPE
Type SPE Tips
Volume (Metric) 200 μL
Quantity 96 Tips/Box
HybridSPE is a registered trademark of Sigma-Aldrich Co. LLC